Ethylenediammonium sulfate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1.2-Diaminoethane sulphate
E0082-25G
E0082-500G
TCE0082-025GEA
38
USD
TCE0082-025G
TCE0082-500G
Ethylenediammonium sulfate ≥98.0% (by titrimetric analysis)
Ethylenediammonium sulphate
CAS Number: 22029-36-3
MDL Number: MFCD00036417
Molecular Formula: C2H8N2
Molecular Weight: 158.17
Purity/Analysis Method:
Formula:
C₂H₈N₂ MW: 158.17 g/mol |
MDL Number:
MFCD00036417 CAS Number: 22029-36-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...