3-Ethoxypropionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
E0053-25ML
E0053-5ML
TCE0053-025MLEA
119.3
USD
TCE0053-025ML
TCE0053-5ML
3-Ethoxypropionic acid ≥98.0% (by GC, titration analysis)
3-Ethoxypropionic acid
CAS Number: 4324-38-3
MDL Number: MFCD00058997
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 219
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: O-Ethylhydracrylic Acid
Formula:
C₅H₁₀O₃ Boiling Pt: 219 °C Density: 1.05 (20 °C) |
MDL Number:
MFCD00058997 CAS Number: 4324-38-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...