Ethanethiol ≥98.0%

Supplier: TCI America
Danger

Synonyms: Ethyl mercaptan, 1-Mercaptoethane

E0036-500ML E0036-25ML
TCE0036-500MLEA 56.96 USD
TCE0036-500ML TCE0036-25ML
Ethanethiol ≥98.0%
Ethanethiol

CAS Number: 75-08-1
MDL Number: MFCD00004887
Molecular Formula: C2H6S
Molecular Weight: 62.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 35
Flash Point (°C): -48
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: Ethanethiol

Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 35 °C
Melting Pt: -148 °C
Density: 0.84 (20 °C)
Flash Pt: -48 °C
Storage Temperature: Ambient
MDL Number: MFCD00004887
CAS Number: 75-08-1
EINECS: 200-837-3
UN: 2363
ADR: 3,I
Merck Index: 12,03771
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34882e21bbd31e06f21192f785a2eb6800df39d2&vid=275722fb1925bacd6b41f94be02c1af92005a805&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR