N,N-Diethyl-m-toluidine ≥99.0% for biochemical research

Supplier: TCI America

Synonyms: 3-Diethylaminotoluene, N,N-Diethyl-3-methylaniline, DMT

D3868-5G D3868-1G
TCD3868-5GEA 80.6 USD
TCD3868-5G TCD3868-1G
N,N-Diethyl-m-toluidine ≥99.0% for biochemical research
N,N-Diethyl-m-toluidine

[for Biochemical Research]
CAS Number: 91-67-8
MDL Number: MFCD00035795
Molecular Formula: C11H17N
Molecular Weight: 163.26
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 104
Specific Gravity (20/20): 0.93


Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 3-Diethylaminotoluene, N,N-Diethyl-3-methylaniline, DMT

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 97
Density: 0.93 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00035795
CAS Number: 91-67-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5b9fcd7d1be5dc1b9da609363e3ab25a4b3e934&vid=43ad54a64295d94b844036dfb169f4c6eb039a4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR