2-Aminocyclohexanol ≥98.0% for synthesis
Supplier: TCI America
|
Danger
|
Synonyms:
1-Amino-2-hydroxycyclohexane, 2-Aminocyclohexanol (cis- and trans- mixture)
A1258-25G
A1258-5G
TCA1258-025GEA
282.5
USD
TCA1258-025G
TCA1258-5G
2-Aminocyclohexanol ≥98.0% for synthesis
2-Aminocyclohexanol
CAS Number: 6850-38-0
MDL Number: MFCD00191368
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1-Amino-2-hydroxycyclohexane
Formula:
C₆H₁₃NO |
MDL Number:
MFCD00191368 CAS Number: 6850-38-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...