Diazolidinyl urea ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 1,3-Bis(hydroxymethyl)-5-[1,3-bis(hydroxymethyl)ureido]hydantoin, 1-(1,3-Bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)-1,3-bis(hydroxymethyl)urea

D3769-250G D3769-25G
TCD3769-250GEA 131.8 USD
TCD3769-250G TCD3769-25G
Diazolidinyl urea ≥98.0% (by total nitrogen basis)
Diazolidinyl urea

CAS Number: 78491-02-8
MDL Number: MFCD03547942
Molecular Formula: C8H14N4O7
Molecular Weight: 278.22
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 1,3-Bis(hydroxymethyl)-5-[1,3-bis(hydroxymethyl)ureido]hydantoin

Formula: C₈H₁₄N₄O₇
MW: 278.22 g/mol
MDL Number: MFCD03547942
CAS Number: 78491-02-8

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR