Diallyl-1,4-cyclohexanedicarboxylate (cis and trans mixture) ≥98.0%
Supplier: TCI America
Synonyms:
1,4-Cyclohexanedicarboxylic Acid Diallyl Ester
D3768-250G
D3768-25G
TCD3768-250GEA
619
USD
TCD3768-250G
TCD3768-25G
Diallyl-1,4-cyclohexanedicarboxylate (cis and trans mixture) ≥98.0%
Diallyl-1,4-cyclohexanedicarboxylate (cis and trans mixture)
CAS Number: 20306-22-3
MDL Number: MFCD00050452
Molecular Formula: C14H20O4
Molecular Weight: 252.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.06
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Cyclohexanedicarboxylic Acid Diallyl Ester
Formula:
C₁₄H₂₀O₄ Density: 1.06 (20 °C) |
MDL Number:
MFCD00050452 CAS Number: 20306-22-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...