2,5-Dimethyl-3-furanthiol ≥97.0%
Supplier: TCI America
Synonyms:
3-Mercapto-2,5-dimethylfuran, 2,5-Dimethylfuran-3-thiol
D3764-1G
D3764-5G
TCD3764-1GEA
57
USD
TCD3764-1G
TCD3764-5G
2,5-Dimethyl-3-furanthiol ≥97.0%
2,5-Dimethyl-3-furanthiol
CAS Number: 55764-23-3
MDL Number: MFCD00801026
Molecular Formula: C6H8OS
Molecular Weight: 128.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 3-Mercapto-2,5-dimethylfuran
Formula:
C₆H₈OS MW: 128.19 g/mol Boiling Pt: 150 °C Density: 1.07 (20 °C) |
MDL Number:
MFCD00801026 CAS Number: 55764-23-3 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...