(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3677-1G D3677-5G
TCD3677-1GEA 65.9 USD
TCD3677-1G TCD3677-5G
(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid

CAS Number: 14590-53-5
MDL Number: MFCD08460918
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 100
Specific Gravity (20/20): 0.99
Specific rotation [a]20/D: 124 deg (C=1, MeOH)

Formula: C₆H₁₀O₂
Boiling Pt: 100 °C
Density: 0.99 (20 °C)
MDL Number: MFCD08460918
CAS Number: 14590-53-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbbeb69140842964d0ca591f8a2c6e25b63ae7f3&vid=cbbeb69140842964d0ca591f8a2c6e25b63ae7f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR