L-Adrenaline bitartrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A0175-25G A0175-1G A0175-5G
TCA0175-025GEA 477 USD
TCA0175-025G TCA0175-001G TCA0175-5G
L-Adrenaline bitartrate ≥98.0% (by HPLC, titration analysis)
L-Adrenaline bitartrate

CAS Number: 51-42-3
MDL Number: MFCD00035077
Molecular Formula: C9H13NO3
Molecular Weight: 333.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 154
Specific rotation [a]20/D: -16.5 deg (C=2, H2O)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: L-Epinephrine Bitartrate, L-Adrenaline Hydrogen Bitartrate, L-Epinephrine Hydrogen Bitartrate

Formula: C₉H₁₃NO₃
Melting Pt: 154 °C
MDL Number: MFCD00035077
CAS Number: 51-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7dfa1bcef2b531706943d0c4a3ff26b6b239e2d&vid=e7dfa1bcef2b531706943d0c4a3ff26b6b239e2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR