(+)-cis-Diltiazem hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, Diltiazem Hydrochloride, (+)-cis-Diltiazem HCl, (+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, (2S,3S)-5-(2-(Dimethylamino)ethyl)-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride
CAS Number: 33286-22-5
MDL Number: MFCD00069252
Molecular Formula: C22H26N2O4S
Molecular Weight: 450.98
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 210
Specific rotation [a]20/D: 118 deg (C=1, H2O)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Antihypertensive Ingredients, Voltage-dependent Calcium Channel (VDCC) Antagonists
Other Product Names / Synonyms: (2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride
Formula:
C₂₂H₂₆N₂O₄S·HCl MW: 450.98 g/mol Melting Pt: 210 °C |
MDL Number:
MFCD00069252 CAS Number: 33286-22-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...