3,5-Dichloro-2,4-difluoroaniline ≥94.0%
Supplier: TCI America
D3637-25G
D3637-5G
TCD3637-25GEA
122.4
USD
TCD3637-25G
TCD3637-5G
3,5-Dichloro-2,4-difluoroaniline ≥94.0%
3,5-Dichloro-2,4-difluoroaniline
CAS Number: 83121-15-7
MDL Number: MFCD00044104
Molecular Formula: C6H3Cl2F2N
Molecular Weight: 197.99
Purity/Analysis Method: >94.0% (GC)
Form: Crystal
Melting point (°C): 75
Formula:
C₆H₃Cl₂F₂N Melting Pt: 75 °C |
MDL Number:
MFCD00044104 CAS Number: 83121-15-7 |
|
Specification Test Results
Purity/Analysis Method | >94.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...