Dibutyl vinylboronate ≥94.0% stabilised

Supplier: TCI America

Synonyms: Vinylboronic Acid Dibutyl Ester, Dibutyl Vinylboronate

D3596-1G D3596-5G
TCD3596-1GEA 92 USD
TCD3596-1G TCD3596-5G
Dibutyl vinylboronate ≥94.0% stabilised
Dibutyl vinylboronate

(stabilized with Phenothiazine)
CAS Number: 6336-45-4
MDL Number: MFCD00050049
Molecular Formula: C10H21BO2
Molecular Weight: 184.09
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 89
Flash Point (°C): 67
Specific Gravity (20/20): 0.84
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Vinylation, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: Vinylboronic Acid Dibutyl Ester

Formula: C₁₀H₂₁BO₂
MW: 184.09 g/mol
Boiling Pt: 89 °C
Density: 0.84 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00050049
CAS Number: 6336-45-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3fc6977b450bacf986a0b1abc58e31687c1aaca&vid=f3fc6977b450bacf986a0b1abc58e31687c1aaca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR