2-Amino-4-chloro-5-nitrophenol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1228-250G A1228-25G
TCA1228-250GEA 344.2 USD
TCA1228-250G TCA1228-025G
2-Amino-4-chloro-5-nitrophenol ≥98.0% (by titrimetric analysis)
2-Amino-4-chloro-5-nitrophenol

CAS Number: 6358-07-2
MDL Number: MFCD00010300
Molecular Formula: C6H5ClN2O3
Molecular Weight: 188.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 220


Other Product Names / Synonyms: 5-Chloro-2-hydroxy-4-nitroaniline

Formula: C₆H₅ClN₂O₃
Melting Pt: 220 °C
MDL Number: MFCD00010300
CAS Number: 6358-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59d122704058b0cfb8e04d66790e16e41ded600&vid=f59d122704058b0cfb8e04d66790e16e41ded600&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR