(+)-Di-p-anisoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (+)-Bis(4-methoxybenzoyl)-D-tartaric Acid, (2S,3S)-2,3-Bis((4-methoxybenzoyl)oxy)succinic acid

D3490-25G D3490-5G
TCD3490-25GEA 170 USD
TCD3490-25G 76219-688
(+)-Di-p-anisoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)
(+)-Di-p-anisoyl-D-tartaric acid

CAS Number: 191605-10-4
MDL Number: MFCD07368366
Molecular Formula: C20H18O10
Molecular Weight: 418.35
Purity/Analysis Method: >98.0% (HPLC,T)
Melting point (°C): 176
Specific rotation [a]20/D: 163 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (+)-Bis(4-methoxybenzoyl)-D-tartaric Acid

Formula: C₂₀H₁₈O₁₀
MW: 418.35 g/mol
Melting Pt: 176 °C
MDL Number: MFCD07368366
CAS Number: 191605-10-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=830389229f3650aad75679429a84d9eb81711c9a&vid=830389229f3650aad75679429a84d9eb81711c9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR