N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine ≥98.0%
Supplier: TCI America
Synonyms:
N-(4-Methyl-2-pentyl)-N'-phenyl-1,4-phenylenediamine, 4-[(4-Methyl-2-pentyl)amino]diphenylamine, N1-(4-Methylpentan-2-yl)-N4-phenylbenzene-1,4-diamine
CAS Number: 793-24-8
Molecular Formula: C18H24N2
Molecular Weight: 268.40
Purity/Analysis Method: >98.0% (GC)
Form: Pellet
Boiling point (°C): 165
Melting point (°C): 51
Flash Point (°C): 204
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 4-[(4-Methyl-2-pentyl)amino]diphenylamine, N-(4-Methyl-2-pentyl)-N'-phenyl-1,4-phenylenediamine
Formula:
C₁₈H₂₄N₂ Boiling Pt: 165 °C Melting Pt: 51 °C Flash Pt: 204 °C |
MDL Number:
MFCD00072248 CAS Number: 793-24-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...