Dihydrojasmonic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-(Carboxymethyl)-2-pentylcyclopentanone, 2-Amyl-3-(carboxymethyl)cyclopentanone, 3-Oxo-2-pentylcyclopentaneacetic Acid, 2-Amyl-3-oxocyclopentaneacetic Acid

D3225-1G D3225-5G
TCD3225-1GEA 265 USD
TCD3225-1G TCD3225-5G
Dihydrojasmonic acid ≥98.0%
Dihydrojasmonic acid

CAS Number: 3572-64-3
Molecular Formula: C12H20O3
Molecular Weight: 212.29
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 1.04
Specific rotation [a]20/D: -83.5 deg (C=0.97, CHCl3)
Storage Temperature: <0°C


Product Categories: Bioscience, Jasmonates
Other Product Names / Synonyms: 2-Amyl-3-(carboxymethyl)cyclopentanone, 2-Amyl-3-oxocyclopentaneacetic Acid, 3-(Carboxymethyl)-2-pentylcyclopentanone, 3-Oxo-2-pentylcyclopentaneacetic Acid

Formula: C₁₂H₂₀O₃
Density: 1.04 (20 °C)
CAS Number: 3572-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48edf41aaab72a4565ace7020397c9eef686e21c&vid=48edf41aaab72a4565ace7020397c9eef686e21c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR