Aluminum glycinate ≥97.0%

Supplier: TCI America

Synonyms: Aminoacetic Acid Dihydroxyaluminum Salt, Dihydroxyaluminum Aminoacetate, Aluminum Glycinate

A1200-25G A1200-500G
TCA1200-025GEA 45 USD
TCA1200-025G TCA1200-500G
Aluminum glycinate ≥97.0%
Aluminum glycinate

CAS Number: 13682-92-3
MDL Number: MFCD00053645
Molecular Formula: C2H6AlNO4
Molecular Weight: 135.05
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Other Product Names / Synonyms: Aminoacetic Acid Dihydroxyaluminum Salt, Dihydroxyaluminum Aminoacetate

Formula: C₂H₆AlNO₄
MW: 135.05 g/mol
MDL Number: MFCD00053645
CAS Number: 13682-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5bdb1dbd83da088442a207739fd41bf8fe321a9&vid=e5bdb1dbd83da088442a207739fd41bf8fe321a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR