Diethyl-3-hydroxyglutarate ≥95.0%

Supplier: TCI America

Synonyms: 3-Hydroxyglutaric Acid Diethyl Ester, Diethyl 3-hydroxyglutarate

D3196-25G D3196-5G
TCD3196-25GEA 174 USD
TCD3196-25G TCD3196-5G
Diethyl-3-hydroxyglutarate ≥95.0%
Diethyl-3-hydroxyglutarate

CAS Number: 32328-03-3
Molecular Formula: C9H16O5
Molecular Weight: 204.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 110
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 3-Hydroxyglutaric Acid Diethyl Ester

Formula: C₉H₁₆O₅
MW: 204.22 g/mol
Boiling Pt: 107 °C
Density: 1.11 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009202
CAS Number: 32328-03-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=119f706ffc2795b332fb35125afc734281ada367&vid=119f706ffc2795b332fb35125afc734281ada367&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR