DL-Adrenaline ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 329-65-7
MDL Number: MFCD00063027
Molecular Formula: C9H13NO3
Molecular Weight: 183.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 197
Storage Temperature: 0-10°C
Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: DL-Epinephrine
Formula:
C₉H₁₃NO₃ Melting Pt: 197 °C |
MDL Number:
MFCD00063027 CAS Number: 329-65-7 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...