1-Acetoxy-2-methoxy-4-(1-propenyl)benzene ≥98.0%
Supplier: TCI America
CAS Number: 93-29-8
MDL Number: MFCD00026984
Molecular Formula: C12H14O3
Molecular Weight: 206.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 283
Melting point (°C): 81
Flash Point (°C): 67
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Acetylisoeugenol
Formula:
C₁₂H₁₄O₃ Boiling Pt: 283 °C Melting Pt: 81 °C Flash Pt: 67 °C |
MDL Number:
MFCD00026984 CAS Number: 93-29-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...