Angelic anhydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1180-1G A1180-5G
TCA1180-001GEA 112.9 USD
TCA1180-001G TCA1180-5G
Angelic anhydride ≥98.0% (by titrimetric analysis)
Angelic anhydride

CAS Number: 94487-74-8
MDL Number: MFCD00198080
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 108
Specific Gravity (20/20): 1.02
Storage Temperature: 0-10°C


Other Product Names / Synonyms: (Z)-2-Methyl-2-butenoic Anhydride, 2-Methylisocrotonic Anhydride

Formula: C₁₀H₁₄O₃
Boiling Pt: 108 °C
Density: 1.02 (20 °C)
MDL Number: MFCD00198080
CAS Number: 94487-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a760cb83b79592ef302276e9a3febd70109686d2&vid=a760cb83b79592ef302276e9a3febd70109686d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR