(R)-(+)-3-Aminopyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (R)-Pyrrolidin-3-amine

A1167-5G A1167-25G
TCA1167-005GEA 293.05 USD
TCA1167-005G TCA1167-025G
(R)-(+)-3-Aminopyrrolidine ≥98.0% (by GC, titration analysis)
(R)-(+)-3-Aminopyrrolidine

CAS Number: 116183-82-5
MDL Number: MFCD00143190
Molecular Formula: C4H10N2
Molecular Weight: 86.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 64
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: 10 deg (C=10, H2O)

Formula: C₄H₁₀N₂
MW: 86.14 g/mol
Density: 0.98 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00143190
CAS Number: 116183-82-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=555dc9d053cf176f12cfae06a11c171717b37833&vid=555dc9d053cf176f12cfae06a11c171717b37833&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR