3,3-Diphenyl-1-propanol ≥98.0%
Supplier: TCI America
D2760-25G
D2760-5G
TCD2760-25GEA
896
USD
TCD2760-25G
TCD2760-5G
3,3-Diphenyl-1-propanol ≥98.0%
3,3-Diphenyl-1-propanol
CAS Number: 20017-67-8
MDL Number: MFCD00002930
Molecular Formula: C15H16O
Molecular Weight: 212.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 110
Specific Gravity (20/20): 1.08
Formula:
C₁₅H₁₆O Boiling Pt: 208 °C Density: 1.08 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00002930 CAS Number: 20017-67-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...