ɑ,ɑ-Dimethyl-γ-butyrolactone ≥97.0%

Supplier: TCI America

Synonyms: Dihydro-3,3-dimethyl-2(3H)-furanone, α,α-Dimethyl-γ-butyrolactone

D2733-25G D2733-5G
TCD2733-25GEA 1325 USD
TCD2733-25G TCD2733-5G
ɑ,ɑ-Dimethyl-γ-butyrolactone ≥97.0%
alpha,alpha-Dimethyl-gamma-butyrolactone

CAS Number: 3709-08-8
MDL Number: MFCD03844788
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 194
Melting point (°C): 6
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Dihydro-3,3-dimethyl-2(3H)-furanone

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 194 °C
Melting Pt: 6 °C
Density: 1.01 (20 °C)
MDL Number: MFCD03844788
CAS Number: 3709-08-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ea32bdada66f6688bce80228711c0413709273e&vid=7ea32bdada66f6688bce80228711c0413709273e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR