Dibutyldichlorosilane ≥94.0%

Supplier: TCI America

D2704-25G D2704-5G
TCD2704-025GEA 344.2 USD
TCD2704-025G TCD2704-005G
Dibutyldichlorosilane ≥94.0%
Dibutyldichlorosilane

CAS Number: 3449-28-3
MDL Number: MFCD00053202
Molecular Formula: C8H18Cl2Si
Molecular Weight: 213.22
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 64
Specific Gravity (20/20): 0.99

Formula: C₈H₁₈Cl₂Si
Boiling Pt: 109 °C
Density: 0.99 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00053202
CAS Number: 3449-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25766739ef33da6143994cdb03610af59e53dfdb&vid=25766739ef33da6143994cdb03610af59e53dfdb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR