2-Anthracenecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Anthroic Acid, 2-Anthracenecarboxylic Acid

A1150-1G A1150-5G
TCA1150-001GEA 212.1 USD
TCA1150-001G TCA1150-005G
2-Anthracenecarboxylic acid ≥98.0% (by GC, titration analysis)
2-Anthracenecarboxylic acid

CAS Number: 613-08-1
MDL Number: MFCD00142865
Molecular Formula: C15H10O2
Molecular Weight: 222.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 289


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 2-Anthroic Acid

Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Melting Pt: 289 °C
MDL Number: MFCD00142865
CAS Number: 613-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3555076c69a174933fd1a2f81b247c71066f68fd&vid=3555076c69a174933fd1a2f81b247c71066f68fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR