(+)-Dipivaloyl-D-tartaric acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(+)-Bis(2,2-dimethylpropionyl)-D-tartaric Acid, (2S,3S)-2,3-Bis(pivaloyloxy)succinic acid, (+)-O,O'-Dipivaloyl-D-tartaric acid
D2702-5G
D2702-1G
TCD2702-005GEA
92
USD
TCD2702-005G
TCD2702-001G
(+)-Dipivaloyl-D-tartaric acid ≥98.0% (by titrimetric analysis)
(+)-Dipivaloyl-D-tartaric acid
CAS Number: 76769-55-6
MDL Number: MFCD00015634
Molecular Formula: C14H22O8
Molecular Weight: 318.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: 24 deg (C=1, Dioxane)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (+)-Bis(2,2-dimethylpropionyl)-D-tartaric Acid
Formula:
C₁₄H₂₂O₈ MW: 318.32 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD00015634 CAS Number: 76769-55-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...