N,N'-Dimethylquinacridone 96,0

Supplier: TCI America

Synonyms: DMQA, 5,12-Dihydro-5,12-dimethylquino[2,3-b]acridine-7,14-dione

D2687-5G D2687-1G
TCD2687-5GEA 235 USD
TCD2687-5G TCD2687-1G
N,N'-Dimethylquinacridone 96,0
N,N'-Dimethylquinacridone

CAS Number: 19205-19-7
MDL Number: MFCD00683078
Molecular Formula: C22H16N2O2
Molecular Weight: 340.38
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Quinacridone Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: 5,12-Dihydro-5,12-dimethylquino[2,3-b]acridine-7,14-dione, DMQA

Formula: C₂₂H₁₆N₂O₂
MW: 340.38 g/mol
MDL Number: MFCD00683078
CAS Number: 19205-19-7

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR