2,4-Dimethylaniline-6-sulfonic acid

Supplier: TCI America

D2680-25G D2680-500G
TCD2680-025GEA 32 USD
TCD2680-025G TCD2680-500G
2,4-Dimethylaniline-6-sulfonic acid
2,4-Dimethylaniline-6-sulfonic acid

CAS Number: 88-22-2
MDL Number: MFCD00035770
Molecular Formula: C8H11NO3S
Molecular Weight: 201.24
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: m-Xylidine-6-sulfonic Acid, 2-Amino-3,5-dimethylbenzenesulfonic Acid

Formula: C₈H₁₁NO₃S
MDL Number: MFCD00035770
CAS Number: 88-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=febbcaa2c59aa48190d3f80b6934a5810d3db8b4&vid=febbcaa2c59aa48190d3f80b6934a5810d3db8b4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR