3,5-Dimethoxybenzyl bromide ≥97.0%
Supplier: TCI America
Synonyms:
1-(Bromomethyl)-3,5-dimethoxybenzene
D2657-5G
D2657-25G
TCD2657-005GEA
91
USD
TCD2657-005G
TCD2657-25G
3,5-Dimethoxybenzyl bromide ≥97.0%
3,5-Dimethoxybenzyl bromide
CAS Number: 877-88-3
MDL Number: MFCD01321368
Molecular Formula: C9H11BrO2
Molecular Weight: 231.09
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 73
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: alpha-Bromo-3,5-dimethoxytoluene
Formula:
C₉H₁₁BrO₂ MW: 231.0864 g/mol Melting Pt: 73 °C |
MDL Number:
MFCD01321368 CAS Number: 877-88-3 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...