Dihydroergotamine mesylate ≥97.0% (by HPLC)
Supplier: TCI America
CAS Number: 6190-39-2
MDL Number: MFCD00058615
Molecular Formula: C33H37N5O5
Molecular Weight: 679.79
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -20 deg (C=0.25, JP Method)
Product Categories: Bioscience, Other Pharmaceutical Ingredients, Indole Alkaloids
Other Product Names / Synonyms: Dihydroergotamine Methanesulfonate
Formula:
C₃₄H₄₁N₅O₈S |
MDL Number:
MFCD00058615 CAS Number: 6190-39-2 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...