Angelic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (Z)-2-Methyl-2-butenoic Acid, 2-Methylisocrotonic Acid, (Z)-2-Methylbut-2-enoic acid

A1136-1G A1136-5G
TCA1136-001GEA 104 USD
TCA1136-001G TCA1136-5G
Angelic acid ≥98.0% (by GC, titration analysis)
Angelic acid

CAS Number: 565-63-9
MDL Number: MFCD00002654
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 44
Storage Temperature: 0-10°C


Product Categories: Bioscience, Hemiterpenes
Other Product Names / Synonyms: (Z)-2-Methyl-2-butenoic Acid, 2-Methylisocrotonic Acid

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 96 °C
Melting Pt: 44 °C
MDL Number: MFCD00002654
CAS Number: 565-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d7ec444579c791aa76f2b177b73f3c506e05fbe&vid=3d7ec444579c791aa76f2b177b73f3c506e05fbe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR