1,4-Dichloro-9,10-anthraquinone ≥98.0%

Supplier: TCI America

Synonyms: 1,4-Dichloroanthracene-9,10-dione

D2518-1G D2518-5G
TCD2518-001GEA 172 USD
TCD2518-001G TCD2518-005G
1,4-Dichloro-9,10-anthraquinone ≥98.0%
1,4-Dichloro-9,10-anthraquinone

CAS Number: 602-25-5
MDL Number: MFCD00192002
Molecular Formula: C14H6Cl2O2
Molecular Weight: 277.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 190

Formula: C₁₄H₆Cl₂O₂
MW: 277.1022 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00192002
CAS Number: 602-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddd4e0f5fca36caaab5722c8c608dee90e4c7ef5&vid=ddd4e0f5fca36caaab5722c8c608dee90e4c7ef5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR