2,4-Diphenyl-4-methyl-1-pentene ≥95.0%
Supplier: TCI America
Synonyms:
4-Methyl-2,4-diphenyl-1-pentene, (4-Methylpent-1-ene-2,4-diyl)dibenzene
D2497-25ML
D2497-500ML
D2497-100ML
TCD2497-025MLEA
21.38
USD
TCD2497-025ML
TCD2497-500ML
TCD2497-100ML
2,4-Diphenyl-4-methyl-1-pentene ≥95.0%
2,4-Diphenyl-4-methyl-1-pentene
CAS Number: 6362-80-7
MDL Number: MFCD00044027
Molecular Formula: C18H20
Molecular Weight: 236.36
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 162
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: 4-Methyl-2,4-diphenyl-1-pentene
Formula:
C₁₈H₂₀ MW: 236.36 g/mol Boiling Pt: 175 °C Density: 0.99 (20 °C) Flash Pt: 162 °C |
MDL Number:
MFCD00044027 CAS Number: 6362-80-7 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...