1,3-Dibromoadamantane ≥97.0%
Supplier: TCI America
D2480-5G
D2480-25G
TCD2480-005GEA
47
USD
TCD2480-005G
TCD2480-25G
1,3-Dibromoadamantane ≥97.0%
1,3-Dibromoadamantane
CAS Number: 876-53-9
MDL Number: MFCD00077218
Molecular Formula: C10H14Br2
Molecular Weight: 294.03
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 111
Formula:
C₁₀H₁₄Br₂ MW: 294.03 g/mol Melting Pt: 111 °C |
MDL Number:
MFCD00077218 CAS Number: 876-53-9 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...