(3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: (2R,3R)-3-((R)-1-((tert-Butyldimethylsilyl)oxy)ethyl)-4-oxoazetidin-2-yl acetate, (3R,4R)-(+)-4-Acetoxy-3-[1-(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone

A1124-5G A1124-25G
TCA1124-005GEA 244.8 USD
TCA1124-005G TCA1124-025G
(3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone ≥98.0% (by total nitrogen basis)
(3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone

CAS Number: 76855-69-1
MDL Number: MFCD00077636
Molecular Formula: C13H25NO4Si
Molecular Weight: 287.43
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 108

Formula: C₁₃H₂₅NO₄Si
MW: 287.43 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00077636
CAS Number: 76855-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d28ad436e9b273603af628e2f5efd2eba75e2c7d&vid=d28ad436e9b273603af628e2f5efd2eba75e2c7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR