trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine ≥98.0%
Supplier: TCI America
Synonyms:
(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, (1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine
CAS Number: 87583-89-9
MDL Number: MFCD00671528
Molecular Formula: C8H18N2
Molecular Weight: 142.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 47
Specific rotation [a]20/D: 145 deg (C=4, CHCl3)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (1S,2S)-(+)-1,2-Bis(methylamino)cyclohexane
Formula:
C8H18N2 MW: 142.25 g/mol Melting Pt: 47 °C |
MDL Number:
MFCD00671528 CAS Number: 87583-89-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...