(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine ≥94.0%

Supplier: TCI America

Synonyms: (1R,2R)-(-)-1,2-Bis(methylamino)cyclohexane, (1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine

D2459-100MG D2459-1G
TCD2459-100MGEA 17 USD
TCD2459-100MG TCD2459-1G
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine ≥94.0%
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine

CAS Number: 68737-65-5
MDL Number: MFCD00671527
Molecular Formula: C8H18N2
Molecular Weight: 142.25
Purity/Analysis Method: >94.0% (GC)
Form: Crystal
Melting point (°C): 45
Specific rotation [a]20/D: -140 deg (C=4, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (1R,2R)-(-)-1,2-Bis(methylamino)cyclohexane

Formula: C₈H₁₈N₂
Melting Pt: 45 °C
MDL Number: MFCD00671527
CAS Number: 68737-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f64442b65c0473fb05c7c2ab8b69ef4a960be1f&vid=9f64442b65c0473fb05c7c2ab8b69ef4a960be1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR