N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
4,4'-Bis[N-phenyl-N-(m-tolyl)amino]biphenyl, N,N'-Diphenyl-N,N'-di(m-tolyl)benzidine, TPD, N,N'-Bis(3-methylphenyl)-N,N'-diphenylbenzidine
CAS Number: 65181-78-4
MDL Number: MFCD00144965
Molecular Formula: C38H32N2
Molecular Weight: 516.69
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 171
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: N,N'-Bis(3-methylphenyl)-N,N'-diphenylbenzidine, 4,4'-Bis[N-phenyl-N-(m-tolyl)amino]biphenyl, TPD
Formula:
C38H32N2 MW: 516.6741 g/mol Melting Pt: 171 °C |
MDL Number:
MFCD00144965 CAS Number: 65181-78-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...