(S,S)-(+)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane ≥97.0% (by titrimetric analysis)
Supplier: TCI America
D2396-1G
D2396-5G
TCD2396-001GEA
182.88
USD
TCD2396-001G
TCD2396-005G
(S,S)-(+)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane ≥97.0% (by titrimetric analysis)
(S,S)-(+)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane
CAS Number: 26549-21-3
MDL Number: MFCD00063598
Molecular Formula: C10H24N2O2
Molecular Weight: 204.31
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Flash Point (°C): 71
Specific Gravity (20/20): 0.90
Specific rotation [a]20/D: 10.5 deg (neat)
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (S,S)-(+)-DDB, (S,S)-(+)-2,3-Dimethoxy-N,N,N',N'-tetramethyl-1,4-butanediamine
Formula:
C₁₀H₂₄N₂O₂ Density: 0.90 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD00063598 CAS Number: 26549-21-3 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...