Cyclohexanemethylamine ≥98.0% for synthesis

Supplier: TCI America

Synonyms: (Cyclohexylmethyl)amine, (Aminomethyl)cyclohexane, Cyclohexylmethanamine

A1114-25ML A1114-100ML
TCA1114-025MLEA 70.1 USD
TCA1114-025ML TCA1114-100ML
Cyclohexanemethylamine ≥98.0% for synthesis
Cyclohexanemethylamine

CAS Number: 3218-02-8
MDL Number: MFCD00001520
Molecular Formula: C7H15N
Molecular Weight: 113.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 163
Flash Point (°C): 48
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Hexahydrobenzylamine, Cyclohexanemethylamine, Cyclohexylmethylamine

Formula: C₇H₁₅N
Boiling Pt: 163 °C
Melting Pt: –8 °C
Density: 0.87 (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00001520
CAS Number: 3218-02-8
EINECS: 221-741-8
UN: 2735
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea3090d209fe2ba6624e1fd67629a02203df2f0&vid=bea3090d209fe2ba6624e1fd67629a02203df2f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR