Dimethyldipalmitylammonium bromide ≥97.0%

Supplier: TCI America

Synonyms: Dihexadecyldimethylammonium Bromide, Dicetyldimonium bromide, N-Hexadecyl-N,N-dimethylhexadecan-1-aminium bromide

D2354-500G D2354-25G
TCD2354-500GEA 974 USD
TCD2354-500G TCD2354-025G
Dimethyldipalmitylammonium bromide ≥97.0%
Dimethyldipalmitylammonium bromide

CAS Number: 70755-47-4
MDL Number: MFCD00191428
Molecular Formula: C34H72BrN
Molecular Weight: 574.86
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Other Product Names / Synonyms: Dihexadecyldimethylammonium Bromide

Formula: C₃₄H₇₂BrN
MW: 574.86 g/mol
MDL Number: MFCD00191428
CAS Number: 70755-47-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eda1873ab3a5d4587869261132597d456801281&vid=6eda1873ab3a5d4587869261132597d456801281&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR