Didecylmethylamine ≥95.0%

Supplier: TCI America

Synonyms: N-Decyl-N-methyldecan-1-amine

D2311-25ML D2311-500ML
TCD2311-025MLEA 27 USD
TCD2311-025ML TCD2311-500ML
Didecylmethylamine ≥95.0%
Didecylmethylamine

CAS Number: 7396-58-9
MDL Number: MFCD00077727
Molecular Formula: C21H45N
Molecular Weight: 311.60
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Melting point (°C): -7
Flash Point (°C): 93
Specific Gravity (20/20): 0.81

Formula: C₂₁H₄₅N
MW: 311.60 g/mol
Boiling Pt: 145 °C
Melting Pt: –7 °C
Density: 0.81 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00077727
CAS Number: 7396-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c56119da46d29778dfdd8048e1e060573a80a346&vid=c56119da46d29778dfdd8048e1e060573a80a346&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR