5,6-Dichloro-2,3-dicyanopyrazine ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
Synonyms:
5,6-Dichloropyrazine-2,3-dicarbonitrile, 5,6-Dichloro-2,3-pyrazinedicarbonitrile
D2271-10G
D2271-25G
TCD2271-010GEA
302
USD
TCD2271-010G
TCD2271-25G
5,6-Dichloro-2,3-dicyanopyrazine ≥98.0% (by GC, total nitrogen)
5,6-Dichloro-2,3-dicyanopyrazine
CAS Number: 56413-95-7
MDL Number: MFCD00191419
Molecular Formula: C6Cl2N4
Molecular Weight: 198.99
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 183
Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 5,6-Dichloro-2,3-pyrazinedicarbonitrile
Formula:
C₆Cl₂N₄ MW: 198.99 g/mol Melting Pt: 183 °C |
MDL Number:
MFCD00191419 CAS Number: 56413-95-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...