2-Methylpentyl acetate ≥98.0%
Supplier: TCI America
Synonyms:
Acetic Acid 2-Methylpentyl Ester
A1079-25ML
A1079-500ML
TCA1079-025MLEA
33
USD
TCA1079-025ML
TCA1079-500ML
2-Methylpentyl acetate ≥98.0%
2-Methylpentyl acetate
CAS Number: 7789-99-3
MDL Number: MFCD00060103
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: Acetic Acid 2-Methylpentyl Ester
Formula:
C₈H₁₆O₂ MW: 144.21 g/mol Boiling Pt: 160 °C Density: 0.87 (20 °C) |
MDL Number:
MFCD00060103 CAS Number: 7789-99-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...