1,4-Dichloro-2-butanol ≥97.0%
Supplier: TCI America
D2194-10G
D2194-25G
D2194-5G
TCD2194-010GEA
339
USD
TCD2194-010G
TCD2194-25G
TCD2194-5G
1,4-Dichloro-2-butanol ≥97.0%
1,4-Dichloro-2-butanol
CAS Number: 2419-74-1
MDL Number: MFCD00191343
Molecular Formula: C4H8Cl2O
Molecular Weight: 143.01
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.29
Formula:
C₄H₈Cl₂O Density: 1.29 (20 °C) |
MDL Number:
MFCD00191343 CAS Number: 2419-74-1 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...