(3R)-(+)-3-(Dimethylamino)pyrrolidine ≥97.0%

Supplier: TCI America

Synonyms: (R)-(+)-3-(Dimethylamino)pyrrolidine, (R)-N,N-Dimethylpyrrolidin-3-amine

D2149-5G D2149-1G
TCD2149-005GEA 285 USD
TCD2149-005G TCD2149-001G
(3R)-(+)-3-(Dimethylamino)pyrrolidine ≥97.0%
(3R)-(+)-3-(Dimethylamino)pyrrolidine

CAS Number: 132958-72-6
MDL Number: MFCD00191347
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 166
Flash Point (°C): 52
Specific Gravity (20/20): 0.90
Specific rotation [a]20/D: 14 deg (C=10, EtOH)

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 166 °C
Density: 0.90 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00191347
CAS Number: 132958-72-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=364d89c9b615aceb8a7801b7b9f249845d1f8c87&vid=364d89c9b615aceb8a7801b7b9f249845d1f8c87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR