Glycerol-ɑ,ɑ'-diallyl ether ≥98.0%

Supplier: TCI America

Synonyms: GDAE, 1,3-Diallyloxy-2-propanol

D2146-500ML D2146-25ML
TCD2146-500MLEA 874 USD
TCD2146-500ML TCD2146-025ML
Glycerol-ɑ,ɑ'-diallyl ether ≥98.0%
Glycerol-alpha,alpha'-diallyl ether

CAS Number: 17018-07-4
MDL Number: MFCD00085245
Molecular Formula: C9H16O3
Molecular Weight: 172.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Specific Gravity (20/20): 0.98


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Diallyloxy-2-propanol, GDAE

Formula: C₉H₁₆O₃
MW: 172.22 g/mol
Boiling Pt: 106 °C
Density: 0.98 (20 °C)
MDL Number: MFCD00085245
CAS Number: 17018-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c490310a3c6e25277962316e404cbcfdb0d5f28&vid=7c490310a3c6e25277962316e404cbcfdb0d5f28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR