1,2-Dichloro-1,1,2,2-tetramethyldisilane ≥95.0%

Supplier: TCI America

D2144-5G D2144-25G
TCD2144-005GEA 97 USD
TCD2144-005G TCD2144-25G
1,2-Dichloro-1,1,2,2-tetramethyldisilane ≥95.0%
1,2-Dichloro-1,1,2,2-tetramethyldisilane

CAS Number: 4342-61-4
MDL Number: MFCD00078528
Molecular Formula: C4H12Cl2Si2
Molecular Weight: 187.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 37
Specific Gravity (20/20): 1.01

Formula: C₄H₁₂Cl₂Si₂
Boiling Pt: 148 °C
Density: 1.01 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00078528
CAS Number: 4342-61-4
EINECS: 224-400-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ce174e1ebbb0b06c1e1e1618dae3cd7ff538777&vid=7ce174e1ebbb0b06c1e1e1618dae3cd7ff538777&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR